missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Epredia™ Consul-Mount™

Designed specifically for use with the Epredia™ Consul™ coverslipper

$315.13 - $327.15

Specifications

Color Clear
Quantity 500 mL
For Use With (Application) For use with Epredia™ Consul™ coverslipper
Form Liquid
View More Specs

Products 2
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
9990440
View Documents Promotion Details
Epredia™
9990440
Mountant N/A
 
9990441
View Documents Promotion Details
Epredia™
9990441
Cytology Media N/A
 
Description

Description

  • Permanent, organic mountant
  • Xylene base
  • Refractive index: 1.495 ±0.005 (dry)
  • Viscosity: Medium (125-200 cps at 24°C)

Specifications

Specifications

Clear
500 mL
For use with Epredia™ Consul™ coverslipper
Liquid
SDS
missing translation for 'documents'

missing translation for 'documents'