Please login to your online account to display your discounted pricing
Thermo Scientific™ Shandon™ 825 Covered Cadaver Carrier
Designed for discrete transport of cadavers
$1,364.58 - $1,585.34
Products
Catalog Number Mfr. No. Material Price Quantity    
106156
thermo scientific™
106156
Naguahyde Each for $1,364.58
23-245-365
thermo scientific™
8253
Fabric Each for $1,585.34
Description & Specifications
  • Two-piece unit includes lightweight canopy and sturdy, elevating gurney
  • Neat, rectangular shape of canopy conceals body for transport
  • Canopy is made from stainless steel with a removable, opaque fabric cover
  • Gurney is constructed from Type 304 stainless steel
  • Table can be raised and lowered via no-slip crank