Thermo Scientific™ Savant SC250EXP SpeedVac Kits
Large-capacity SpeedVac kits—everything you need to effectively concentrate high boiling point solvents
Manufacturer:  Thermo Scientific™ SC250EXPA-115
Catalog No. 11-716-811
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Dual vacuum control on our SPD250EXP provides flexibility to use many sample types.
  • Four IR heat lamps for drying high boiling point solvents such as DMSO and DMF
  • Dual vacuum control of RAMP and LEVEL eliminates samples from bumping and/or freezing, providing optimization for various sample types
  • Kalrez™ and PTFE-coated materials along the entire vapor path for maximum chemical resistance and durabilityi
  • Dual valve mechanism for improved durability and exhausting
  • Comes complete with enlarged vacuum port, extra wide tubing and new extra large mouth glass condensation flask
  • Heated glass lid reduces drying times and prevents condensation
  • Digital display of parameters (time, temperture and vacuum)
  • Standard glass safety lids
Solvent and Solvent Combinations - SC250P1
  • Acetic Acid
  • Acetone
  • Ammonium Hydroxide
  • Formic Acid
  • Hydrochloric Acid
  • TFA
  • Methylane Chloride and Water
  • Drug Metabolites in SPE/Urine
  • DCM and Hepatane
  • Methylene Chloride and MEOH
  • THF/Pyridine (95:5)
  • DMSO and Methanol
Solvent and Solvent Combinations - SC250P2
  • Chloroform
  • DMSO
  • DMF
  • Ethyl Acetate
  • Hexane
  • Toluene
  • Methanol and Chloroform
SC250P1
  • Solid phase extractions
  • Drying lipid extracts
  • Flash chromatography fractions
  • Solid phase synthesis cleavage solutions such as protein hydrolyzates and evaporating synthesis solids
SC250P2
  • Complete management of evaporating dissolution solvents
Specifications
Specifications
115 V
60 Hz
115 V 60 Hz