Please login to your online account to display your discounted pricing

Thermo Scientific™ Savant™ DNA SpeedVac™ Concentrator Kits

Compact, fully integrated systems designed specifically for DNA and RNA applications

Specifications

Dimensions (L x W x H) 26 x 25 x 15 in. (66 x 62 x 38cm)
Includes One RH64-11 rotor and two GCF400 Glass condensation flask
Max. Vacuum 9mbar
Shipping Weight (English) 175 lb.
Shipping Weight (Metric) 79kg
View More Specs
Includes:  DNA120
  • DNA concentrator with integrated pump
  • RD36 rotor for 36 x 1.6mL microtubes
DNA120-OP
  • DNA concentrator with integrated pump and glass cover
  • RD36 rotor for 36 x 1.5mL microtubes
  • ANT100 chemical trap
  • ANS121 ammonium neutralizing solution
Products
Catalog Number Mfr. No. Capacity (Metric) Hertz Voltage Price Quantity    

36-792-1

 
thermo scientific™
ISS110115
4000mL 60Hz 115V CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Dimensions (L x W x H) 26 x 25 x 15 in. (66 x 62 x 38cm)
Includes One RH64-11 rotor and two GCF400 Glass condensation flask
Max. Vacuum 9mbar
Shipping Weight (English) 175 lb.
Shipping Weight (Metric) 79kg
Temperature Range (Metric) -55°C
Material PTFE
Wattage 1150w
Weight (English) 135 lb.
Weight (Metric) 61kg

The DNA-series SpeedVac vacuum concentrators deliver rapid and efficient concentration and drying of DNA/RNA samples, nucleic acids, PCR preps and synthetic oligos. These easy-to-use, quiet integrated systems are designed to accommodate microcentrifuge tubes or shallow-well microplates. A built-in, oil-free PTFE pump produces the vacuum required for rapid drying of samples with clean vacuum and no regular maintenance required. Choice of three models to handle low-volume sample preps as well as higher-volume processing requirements. Units are easy to use and install with no additional tubing or pumps required.

Operating Features

  • Consistent performance: designed to ensure consistent drying times and reproducible results
  • Simplicity: single program supports all DNA/RNA applications
  • User-friendly interface: with temperature/time settings
  • Three drying rates: ambient, ∽43°C, ∽65°C
  • Dual digital timers: independent control of heat/run times
Sample Protection
  • Dry at ambient temperatures or at very low heat to avoid sample denaturing
  • Oil-free diaphragm pump provides gentle transition from atmosphere to ultimate vacuum setting with no bumping
Safety Features
  • PTFE-coated chamber resists chemicals and cleans easily
  • Latched lid ensures user safety during operation
  • Unit and pump are virtually maintenance-free
  • Optional chemical traps and cartridges trap radioactivity and ammonia

Solvents and Solvent Combinations for DNA120

  • PCR buffers (aqueous)
  • Water
  • Ethanol and water
Solvents and Solvent Combinations for DNA-120-OP
  • Ammonium hydroxide
  • Isopropanel
  • PCR buffers (aqueous)
Solvents and Solvent Combinations for ISS-110
  • PCR buffers (aqueous)
  • Ethanol and water
  • Isopropanol

DNA 120 SpeedVac Kit

  • Corrosion-resistant, PTFE coated chamber provides years of lasting service
  • Built-in, oil free vacuum pump provides maintenance-free operation
  • Integrated and compact design
  • Extremely quiet operation
  • Choice of three drying rates
  • Dual digital timers for independent control of heat and run times
DNA120OP SpeedVac Kitfor oligos
  • All the features of the DNA 120
  • Plus external post trap to neutralize ammonium hydroxide, chemically resistant glass cover to contain solvents
High capacity DNA SpeedVac Kit ISS110
  • Built-in ETFE coated vacuum pump providing years of lasting service
  • Four liters, -50°C refrigerated vapor trap (integrated)
  • Choice of three drying rates

DNA120

  • DNA/RNA preparation prior to analysis by electrophoresis or microarrays
  • Labeling cDNA with flourescent dyes
  • Drying PCR samples in aqueous buffers
DNA120-OP
  • Drying synthetic oligonucleotides in ammonium hydroxide

CE (230V only)

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.