Thermo Scientific™ Rotors for Savant™ DNA SpeedVac™ Concentrators
Accessory Rotors
Manufacturer:  Thermo Scientific™ RH2012
 View more versions of this product
Catalog No. 36-073-2
Description
Description
Easy to use circular rotors with different tube hole sizes to accommodate different sample sizes. These rotors require no additional accessories and fit directly into your concentrator.
Specifications
Specifications
20 Tube
Savant™ DNA SpeedVac™ concentrator rotor
Glass And Plastic Tube
20.0