Please login to your online account to display your discounted pricing
Thermo Scientific™ Rotors for Savant™ DNA SpeedVac™ Concentrators
Accessory Rotors
$1,239.68 - $1,239.68
Specifications
For Use With (Equipment) Savant™ DNA SpeedVac™ concentrator rotor
View More Specs
Products
Catalog Number Mfr. No. Total Count Product Type Price Quantity    
36-073-2
thermo scientific™
RH2012
20.0 Glass And Plastic Tube Each for $1,239.68
Description & Specifications
Specifications
For Use With (Equipment) Savant™ DNA SpeedVac™ concentrator rotor
Easy to use circular rotors with different tube hole sizes to accommodate different sample sizes. These rotors require no additional accessories and fit directly into your concentrator.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.