Thermo Scientific™ Remel™ Bordet Gengou Agar (with blood)
With blood; For cultivation and isolation of Bordetella pertussis and other Bordetella spp.
$179.01 - $179.01
Specifications
Certifications/Compliance USDA, APHIS
For Use With (Application) Cultivation of fastidious microorganisms and visualization of hemolytic reactions
Media Type Bordet Gengou Agar with Blood
Manufacturing Origin United States
View More Specs
Disclaimers:
Part No. R09192 required addition of sheep, rabbit or horse blood. Part No. R110810 not intended for IVD use.
Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
R01240 Thermo Scientific™
RE01240
Plate 10/Pack Pack for $179.01
Specifications
Description
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
USDA, APHIS
Cultivation of fastidious microorganisms and visualization of hemolytic reactions
Bordet Gengou Agar with Blood
United States