Thermo Scientific™ PocketTips™
Nanoliter dispensing pipette tips with an internally molded capillary pocket that holds 50 to 500nL
$1,055.03 - $1,055.03
Specifications
Tip Style Automation
View More Specs
Products 1
Catalog Number Mfr. No. For Use With (Equipment) Volume (Metric) Price Quantity  
Catalog Number Mfr. No. For Use With (Equipment) Volume (Metric) Price Quantity  
14-387-380
View Documents
Thermo Scientific™
U200250
Hamilton™ MICROLAB™ STAR 250 nL
Case of 4800 Each for $1,055.03
Description
Description
A broad range of pocket volumes are compatible with many industry standard 96- and 384-channel automated liquid handling platforms to allow existing equipment to implement new innovative pipetting procedures.
  • Transfer samples directly to the assay plate
  • Combine sample and assay material with soft in-tip mixing—thorough mixing without damaging sensitive material
  • Consistent accuracy and precision; coefficient of variation of less than 6% can be achieved for any pocket size
  • Color: Clear
Specifications
Specifications
Automation
SDS