Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-Visfatin (human) Recombinant, Thermo Scientific™

Visfatin (Hu) Recombinant protein RP-75759 from Thermo Scientific™ specifically detects Visfatin (Hu) in Not Applicable samples, and it is validated for Control and WB

$361.05 - $4,038.25

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human Visfatin
Recombinant Yes
Source E. coli
View More Specs

 Disclaimers

in vitro research only.

Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

PIRP75757

 
pierce biotechnology inc
RP75757
10μg purified protein (0.88mg/mL) in Laemmli and no preservative 10μg Each for $361.05

PIRP75759

 
pierce biotechnology inc
RP75759
1mg purified protein in tris HCl with 50% glycerol and no preservative; pH 7.9 1mg Each for $4,038.25
Description & Specifications

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human Visfatin
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

Visfatin (Hu) Recombinant protein RP-75759 from Thermo Scientific™ specifically detects Visfatin (Hu) in Not Applicable samples, and it is validated for Control and WB


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 1 mg purified Protein in tris HCl, with 50% glycerol and no preservative; 7.9

RP-75759 detects Visfatin (human) in Not Applicable samples. RP-75759 has been successfully used in Control and Western Blot procedures. RP-75759 The immunogen is N/A.