Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-VEGF Receptor 1 (human) Recombinant, Thermo Scientific™

VEGF Receptor 1 (Hu) Recombinant protein RP-75750 from Thermo Scientific™ specifically detects VEGF Receptor 1 (Hu) in Not Applicable samples, and is validated for Control and WB

$272.60 - $4,038.25

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human VEGF Receptor 1
Recombinant Yes
Source E. coli
View More Specs

 Disclaimers

in vitro research only.

Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

PIRP75748

 
pierce biotechnology inc
RP75748
10μg purified protein (0.11mg/mL) in 0.025M acetic acid with 50% glycerol and no preservative; pH 4.8 10μg Each for $272.60

PIRP75749

 
pierce biotechnology inc
RP75749
100μg purified protein in sodium acetate with 50% glycerol and no preservative; pH 4.8 100μg Each for $870.00

PIRP75750

 
pierce biotechnology inc
RP75750
1mg purified protein in 0.025M acetic acid with 50% glycerol and no preservative; pH 4.8 1mg Each for $4,038.25
Description & Specifications

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human VEGF Receptor 1
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

VEGF Receptor 1 (Hu) Recombinant protein RP-75750 from Thermo Scientific™ specifically detects VEGF Receptor 1 (Hu) in Not Applicable samples, and is validated for Control and WB


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 1 mg purified Protein in 0.025M acetic acid, with 50% glycerol and no preservative; 4.8

RP-75750 detects VEGF Receptor 1 (human) in Not Applicable samples. RP-75750 has been successfully used in Control and Western Blot procedures. RP-75750 The immunogen is N/A.