Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-VCAM1 (human) Recombinant, Thermo Scientific™

VCAM1 (human) Recombinant protein RP-75753 from Thermo Scientific™ specifically detects VCAM1 (human) in Not Applicable samples, and it is validated for Control and Western Blot

$3,770.00 - $3,770.00

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human VCAM1
Recombinant Yes
Source E. coli
View More Specs

 Disclaimers

in vitro research only.

Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

PIRP75753

 
pierce biotechnology inc
RP75753
1mg purified protein in PBS with 50% glycerol and no preservative 1mg Each for $3,770.00
Description & Specifications

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human VCAM1
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

VCAM1 (human) Recombinant protein RP-75753 from Thermo Scientific™ specifically detects VCAM1 (human) in Not Applicable samples, and it is validated for Control and Western Blot


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 1 mg purified Protein in PBS, with 50% glycerol and no preservative;

RP-75753 detects VCAM1 (human) in Not Applicable samples. RP-75753 has been successfully used in Control and Western Blot procedures. RP-75753 The immunogen is N/A.