Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-VCAM1 Fragment (human) Recombinant, Thermo Scientific™

VCAM1 Fragment (Hu) Recombinant protein RP-75756 from Thermo Scientific™ specifically detects VCAM1 Fragment (Hu) in Not Applicable samples, and is validated for Control and WB

$288.55 - $3,770.00

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human VCAM1 Fragment
Recombinant Yes
Source E. coli
View More Specs

 Disclaimers

in vitro research only.

Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

PIRP75754

 
pierce biotechnology inc
RP75754
10μg purified protein in PBS with 50% glycerol and no preservative 10μg Each for $288.55

PIRP75756

 
pierce biotechnology inc
RP75756
1mg purified protein in PBS with 50% glycerol and no preservative 1mg Each for $3,770.00

PIRP75755

 
pierce biotechnology inc
RP75755
100μg purified protein in PBS with 50% glycerol and no preservative 100μg Each for $797.50
Description & Specifications

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human VCAM1 Fragment
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

VCAM1 Fragment (Hu) Recombinant protein RP-75756 from Thermo Scientific™ specifically detects VCAM1 Fragment (Hu) in Not Applicable samples, and is validated for Control and WB


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 1 mg purified Protein in PBS, with 50% glycerol and no preservative;

RP-75756 detects VCAM1 Fragment (human) in Not Applicable samples. RP-75756 has been successfully used in Control and Western Blot procedures. RP-75756 The immunogen is N/A.