Please login to your online account to display your discounted pricing

anti-Vascular Endothelial Growth Factor (human) Recombinant, Thermo Scientific™


Vascular Endothelial Growth Factor (Hu) Recombinant protein RP-75747 detects Vascular Endothelial Growth Factor (Hu) in Not Applicable samples, and is validated for Control and WB

Manufacturer: pierce biotechnology inc  RP75747

 View more versions of this product

 Disclaimers

in vitro research only.

Catalog No. PIRP75747

  • / Each
 

Description & Specifications

Specifications

Formulation 1mg purified protein in 20mM tris HCl with 50% glycerol, 5mM EDTA and no preservative
For Use With (Application) Control, Western Blot
Host Species E. Coli
Conjugate Unlabeled
Format Purified
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Immunogen N/A
Source E. coli
Name Human Vascular Endothelial Growth Factor
Recombinant Yes
Cross Reactivity Human
Quantity 1mg

Vascular Endothelial Growth Factor (Hu) Recombinant protein RP-75747 detects Vascular Endothelial Growth Factor (Hu) in Not Applicable samples, and is validated for Control and WB


Host Species: E. Coli
Formulation: 1mg purified protein in 20mM tris HCl with 50% glycerol, 5mM EDTA and no preservative
RP-75747 contains purified recombinant human VeGF protein with an N-terminal hexahistidine tag. RP-75747 may be used as a positive control in many immunological assays.