Please login to your online account to display your discounted pricing

anti-Vascular Endothelial Growth Factor (human) Recombinant, Thermo Scientific™


Vascular Endothelial Growth Factor (Hu) Recombinant protein RP-75747 detects Vascular Endothelial Growth Factor (Hu) in Not Applicable samples, and is validated for Control and WB

Manufacturer: pierce biotechnology inc  RP75745

 View more versions of this product

 Disclaimers

in vitro research only.

Catalog No. PIRP75745

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
For Use With (Application) Control, Western Blot
Formulation 10μg purified protein in 20mM tris HCl with 50% glycerol, 5mM EDTA and no preservative; pH 8
Host Species E. Coli
Immunogen N/A
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Format Purified
Name Human Vascular Endothelial Growth Factor 121
pH Range 8
Recombinant Yes
Source E. coli
Quantity 10μg
Cross Reactivity Human

Vascular Endothelial Growth Factor (Hu) Recombinant protein RP-75747 detects Vascular Endothelial Growth Factor (Hu) in Not Applicable samples, and is validated for Control and WB


Host Species: E. Coli
Formulation: 10μg purified protein in 20mM tris HCl with 50% glycerol, 5mM EDTA and no preservative; pH 8
RP-75745 contains purified recombinant human VeGF protein with an N-terminal hexahistidine tag. RP-75745 may be used as a positive control in many immunological assays.