Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-Vascular Endothelial Growth Factor (human) Recombinant, Thermo Scientific™

Vascular Endothelial Growth Factor (Hu) Recombinant protein RP-75747 detects Vascular Endothelial Growth Factor (Hu) in Not Applicable samples, and is validated for Control and WB

$272.60 - $3,770.00

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
View More Specs

 Disclaimers

in vitro research only.

Products
Catalog Number Mfr. No. Formulation Name Quantity Price Quantity    

PIRP75745

 
pierce biotechnology inc
RP75745
10μg purified protein in 20mM tris HCl with 50% glycerol, 5mM EDTA and no preservative; pH 8 Human Vascular Endothelial Growth Factor 121 10μg Each for $272.60

PIRP75746

 
pierce biotechnology inc
RP75746
100μg purified protein in 20mM tris HCl with 50% glycerol, 5mM EDTA and no preservative; pH 8 Human Vascular Endothelial Growth Factor 121 100μg Each for $870.00

PIRP75747

 
pierce biotechnology inc
RP75747
1mg purified protein in 20mM tris HCl with 50% glycerol, 5mM EDTA and no preservative Human Vascular Endothelial Growth Factor 1mg Each for $3,770.00
Description & Specifications

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

Vascular Endothelial Growth Factor (Hu) Recombinant protein RP-75747 detects Vascular Endothelial Growth Factor (Hu) in Not Applicable samples, and is validated for Control and WB


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 1 mg purified Protein in PBS, with 50% glycerol and no preservative;

RP-75747 detects Vascular Endothelial Growth Factor (human) in Not Applicable samples. RP-75747 has been successfully used in Control and Western Blot procedures. RP-75747 The immunogen is N/A.