Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-TREM1 (human) Recombinant, Thermo Scientific™

TREM1 (human) Recombinant protein RP-75732 from Thermo Scientific™ specifically detects TREM1 (human) in Not Applicable samples, and it is validated for Control and Western Blot

$800.40 - $3,770.00

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Molecular Weight 24.76
Name Human TREM1 Recombinant Protein
Recombinant Yes
View More Specs

 Disclaimers

in vitro research only.

Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

PIRP75731

 
pierce biotechnology inc
RP75731
100μg purified protein (0.23mg/mL) in TBS with 50% glycerol and no preservative; pH 8 100μg Each for $800.40

PIRP75732

 
pierce biotechnology inc
RP75732
1mg purified protein in PBS with 50% glycerol and no preservative 1mg Each for $3,770.00
Description & Specifications

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Molecular Weight 24.76
Name Human TREM1 Recombinant Protein
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

TREM1 (human) Recombinant protein RP-75732 from Thermo Scientific™ specifically detects TREM1 (human) in Not Applicable samples, and it is validated for Control and Western Blot


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 1 mg purified Protein in PBS, with 50% glycerol and no preservative;
Synonyms: Soluble Triggering Receptor Expressed on Myeloid Cells-1

RP-75732 detects TREM1 (human) in Not Applicable samples. RP-75732 has been successfully used in Control and Western Blot procedures. RP-75732 The immunogen is N/A.