Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-TIMP1 (human) Recombinant, Thermo Scientific™

TIMP1 (human) Recombinant protein RP-75735 from Thermo Scientific™ specifically detects TIMP1 (human) in Not Applicable samples, and it is validated for Control and Western Blot

$910.60 - $3,770.00

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human TIMP1 Recombinant Protein
Recombinant Yes
Source E. coli
View More Specs

 Disclaimers

in vitro research only.

Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

PIRP75735

 
pierce biotechnology inc
RP75735
1mg purified protein in PBS with 50% glycerol and no preservative 1mg Each for $3,770.00

PIRP75734

 
pierce biotechnology inc
RP75734
100μg purified protein (0.31mg/mL) in 0.025M acetic acid with 50% glycerol and no preservative; pH 4.8 100μg Each for $910.60
Description & Specifications

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human TIMP1 Recombinant Protein
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

TIMP1 (human) Recombinant protein RP-75735 from Thermo Scientific™ specifically detects TIMP1 (human) in Not Applicable samples, and it is validated for Control and Western Blot


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 1 mg purified Protein in PBS, with 50% glycerol and no preservative;
Synonyms: Tissue Inhibitor of Metalloproteinase 1,TIMP1

RP-75735 detects TIMP1 (human) in Not Applicable samples. RP-75735 has been successfully used in Control and Western Blot procedures. RP-75735 The immunogen is N/A.