Please login to your online account to display your discounted pricing

anti-soluble TNF Receptor II (human) Recombinant, Thermo Scientific™


soluble TNF Receptor II (Hu) Recombinant protein RP-75729 detects soluble TNF Receptor II (Hu) in Not Applicable samples, and is validated for Control and WB

Manufacturer: pierce biotechnology inc  RP75727

 View more versions of this product

 Disclaimers

in vitro research only.

Catalog No. PIRP75727

  • / Each
 

Description & Specifications

Specifications

Formulation 10μg purified protein (0.3mg/mL) in tris HCl with 50% glycerol, 5mM EDTA and no preservative; pH 8
For Use With (Application) Control, Western Blot
Host Species E. Coli
Conjugate Unlabeled
Format Purified
Immunogen N/A
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Source E. coli
Name Human Soluble TNF Receptor II Recombinant Protein
Recombinant Yes
Cross Reactivity Human
Concentration 0.3mg/mL
pH Range 8
Quantity 10μg

soluble TNF Receptor II (Hu) Recombinant protein RP-75729 detects soluble TNF Receptor II (Hu) in Not Applicable samples, and is validated for Control and WB


Host Species: E. Coli
Formulation: 10μg purified protein (0.3mg/mL) in tris HCl with 50% glycerol, 5mM EDTA and no preservative; pH 8
RP-75727 contains purified recombinant, soluble human TNF Receptor-2 protein with an N-terminal hexahistidine tag. RP-75727 may be used as a positive control in many immunological assays.