Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-soluble TNF Receptor II (human) Recombinant, Thermo Scientific™

soluble TNF Receptor II (Hu) Recombinant protein RP-75729 detects soluble TNF Receptor II (Hu) in Not Applicable samples, and is validated for Control and WB

$288.55 - $3,770.00

Specifications

Concentration 0.3mg/mL
Cross Reactivity Human
Formulation 1mg purified protein in PBS with 50% glycerol and no preservative
For Use With (Application) Control, Western Blot
Name Human Soluble TNF Receptor II Recombinant Protein
View More Specs

 Disclaimers

in vitro research only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

PIRP75727

 
pierce biotechnology inc
RP75727
10μg Each for $288.55

PIRP75728

 
pierce biotechnology inc
RP75728
100μg Each for $797.50

PIRP75729

 
pierce biotechnology inc
RP75729
1mg Each for $3,770.00
Description & Specifications

Specifications

Concentration 0.3mg/mL
Cross Reactivity Human
Formulation 1mg purified protein in PBS with 50% glycerol and no preservative
For Use With (Application) Control, Western Blot
Name Human Soluble TNF Receptor II Recombinant Protein
pH Range 8
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

soluble TNF Receptor II (Hu) Recombinant protein RP-75729 detects soluble TNF Receptor II (Hu) in Not Applicable samples, and is validated for Control and WB


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 1 mg purified Protein in PBS, with 50% glycerol and no preservative;

RP-75729 detects soluble TNF Receptor II (human) in Not Applicable samples. RP-75729 has been successfully used in Control and Western Blot procedures. RP-75729 The immunogen is N/A.