Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-soluble TNF Receptor I (human) Recombinant, Thermo Scientific™

soluble TNF Receptor I (Hu) Recombinant protein RP-75726 detects soluble TNF Receptor I (Hu) in Not Applicable samples, and is validated for Control and WB

$272.60 - $3,770.00

Specifications

Concentration 0.1mg/mL
Cross Reactivity Human
Formulation 1mg purified protein in PBS with 50% glycerol and no preservative
For Use With (Application) Control, Western Blot
Name Human Soluble TNF Receptor I Recombinant Protein
View More Specs

 Disclaimers

in vitro research only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

PIRP75725

 
pierce biotechnology inc
RP75725
100μg Each for $785.90

PIRP75726

 
pierce biotechnology inc
RP75726
1mg Each for $3,770.00

PIRP75724

 
pierce biotechnology inc
RP75724
10μg Each for $272.60
Description & Specifications

Specifications

Concentration 0.1mg/mL
Cross Reactivity Human
Formulation 1mg purified protein in PBS with 50% glycerol and no preservative
For Use With (Application) Control, Western Blot
Name Human Soluble TNF Receptor I Recombinant Protein
pH Range 8
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

soluble TNF Receptor I (Hu) Recombinant protein RP-75726 detects soluble TNF Receptor I (Hu) in Not Applicable samples, and is validated for Control and WB


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 1 mg purified Protein in PBS, with 50% glycerol and no preservative;

RP-75726 detects soluble TNF Receptor I (human) in Not Applicable samples. RP-75726 has been successfully used in Control and Western Blot procedures. RP-75726 The immunogen is N/A.