Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-S100A11 (human) Recombinant, Thermo Scientific™

S100A11 (Hu) Recombinant protein RP-75720 from Thermo Scientific™ specifically detects S100A11 (Hu) in Not Applicable samples, and it is validated for Control and WB

$303.05 - $3,770.00

Specifications

Concentration 0.05mg/mL
Cross Reactivity Human
Formulation 1mg purified protein in PBS with 50% glycerol and no preservative
For Use With (Application) Control, Western Blot
Molecular Weight ∽17.1
View More Specs

 Disclaimers

in vitro research only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

PIRP75719

 
pierce biotechnology inc
RP75719
100μg Each for $723.55

PIRP75718

 
pierce biotechnology inc
RP75718
10μg Each for $303.05

PIRP75720

 
pierce biotechnology inc
RP75720
1mg Each for $3,770.00
Description & Specifications

Specifications

Concentration 0.05mg/mL
Cross Reactivity Human
Formulation 1mg purified protein in PBS with 50% glycerol and no preservative
For Use With (Application) Control, Western Blot
Molecular Weight ∽17.1
Name Human S100A11 Recombinant Protein
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

S100A11 (Hu) Recombinant protein RP-75720 from Thermo Scientific™ specifically detects S100A11 (Hu) in Not Applicable samples, and it is validated for Control and WB


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 1 mg purified Protein in PBS, with 50% glycerol and no preservative;

RP-75720 detects S100A11 (human) in Not Applicable samples. RP-75720 has been successfully used in Control and Western Blot procedures. RP-75720 The immunogen is N/A.