Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-S100A10 (human) Recombinant, Thermo Scientific™

S100A10 (Hu) Recombinant protein RP-75717 from Thermo Scientific™ specifically detects S100A10 (Hu) in Not Applicable samples, and it is validated for Control and WB

$288.55 - $3,770.00

Specifications

Cross Reactivity Human
Formulation 1mg purified protein in PBS with 50% glycerol and no preservative
For Use With (Application) Control, Western Blot
Name Human S100A10 Recombinant Protein
Recombinant Yes
View More Specs

 Disclaimers

in vitro research only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

PIRP75717

 
pierce biotechnology inc
RP75717
1mg Each for $3,770.00

PIRP75715

 
pierce biotechnology inc
RP75715
10μg Each for $288.55
Description & Specifications

Specifications

Cross Reactivity Human
Formulation 1mg purified protein in PBS with 50% glycerol and no preservative
For Use With (Application) Control, Western Blot
Name Human S100A10 Recombinant Protein
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

S100A10 (Hu) Recombinant protein RP-75717 from Thermo Scientific™ specifically detects S100A10 (Hu) in Not Applicable samples, and it is validated for Control and WB


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 1 mg purified Protein in PBS, with 50% glycerol and no preservative;

RP-75717 detects S100A10 (human) in Not Applicable samples. RP-75717 has been successfully used in Control and Western Blot procedures. RP-75717 The immunogen is N/A.