Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-S100 beta Chain (human) Recombinant, Thermo Scientific™

S100 beta Chain (Hu) Recombinant protein RP-75723 from Thermo Scientific™ specifically detects S100 beta Chain (Hu) in Not Applicable samples, and is validated for Control and WB

$326.25 - $3,770.00

Specifications

Concentration 0.1mg/mL
Cross Reactivity Human
Formulation 1mg purified protein in PBS with 50% glycerol and no preservative
For Use With (Application) Control, Western Blot
Molecular Weight ∽15
View More Specs

 Disclaimers

in vitro research only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

PIRP75721

 
pierce biotechnology inc
RP75721
10μg Each for $326.25

PIRP75723

 
pierce biotechnology inc
RP75723
1mg Each for $3,770.00

PIRP75722

 
pierce biotechnology inc
RP75722
100μg Each for $688.75
Description & Specifications

Specifications

Concentration 0.1mg/mL
Cross Reactivity Human
Formulation 1mg purified protein in PBS with 50% glycerol and no preservative
For Use With (Application) Control, Western Blot
Molecular Weight ∽15
Name Human S100 beta Chain Recombinant Protein
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

S100 beta Chain (Hu) Recombinant protein RP-75723 from Thermo Scientific™ specifically detects S100 beta Chain (Hu) in Not Applicable samples, and is validated for Control and WB


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 1 mg purified Protein in PBS, with 50% glycerol and no preservative;

RP-75723 detects S100 beta Chain (human) in Not Applicable samples. RP-75723 has been successfully used in Control and Western Blot procedures. RP-75723 The immunogen is N/A.