Please login to your online account to display your discounted pricing

RuvC Recombinant Protein, Invitrogen™


RuvC Recombinant Protein, RP-4911, from Invitrogen™

Manufacturer: pierce biotechnology inc  RP4911

Catalog No. ARC4911

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Control, Western Blot
Formulation 100μg purified protein (13mg/mL) in TBS with 50% glycerol, 5mM beta mercaptoethanol, 2mM EDTA and no preservative; pH 7.5
Conjugate Unlabeled
Host Species E. Coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Format Purified
Molecular Weight ∽19
pH Range 7.5
Source E. coli
Recombinant Yes
Purification Method Chromatography
Name RuvC Recombinant Protein
Quantity 100μg

RuvC Recombinant Protein, RP-4911, from Invitrogen.

E. coli RuvC protein is a structurally specific endonuclease which binds specifically to the Holliday structure, an intermediate of recombination, at the late stage of homologous recombination and recombination repair and introduces a nick in the symmetrical point of the Holliday junction cleaving and resolving the recombinant. The product is a recombinant protein abundantly expressed by E. coli and purified by methods such as chromatography. Its molecular weight is 19 kDa and forms a dimer in liquid and in binding state with the Holliday structure.
Host Species: E. Coli
Formulation: 100μg purified protein (13mg/mL) in TBS with 50% glycerol, 5mM beta mercaptoethanol, 2mM EDTA and no preservative; pH 7.5
RP-4911 can be used as a positive control in Western blotting, as well as in studies in studies on homologous recombination mechanism. RP-4911 calculated molecular weight is ∽19 kDa. Purity is 90%.