Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-Retinol Binding Protein (human) Recombinant, Thermo Scientific™

Retinol Binding Protein (Hu) Recombinant protein RP-75714 detects Retinol Binding Protein (Hu) in Not Applicable samples, and is validated for Control and WB

$361.05 - $3,770.00

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
View More Specs

 Disclaimers

in vitro research only.

Products
Catalog Number Mfr. No. Formulation Name Quantity Price Quantity    

PIRP75712

 
pierce biotechnology inc
RP75712
10μg purified protein (0.05mg/mL) in Laemmli and no preservative; pH 6.8 Human Retinol Binding Recombinant Protein with Amino-terminal 6x Histidine Tag 10μg Each for $361.05

PIRP75714

 
pierce biotechnology inc
RP75714
1mg purified protein in PBS with 50% glycerol and no preservative Human Retinol Binding Recombinant Protein 1mg Each for $3,770.00

PIRP75713

 
pierce biotechnology inc
RP75713
100μg purified protein in PBS with 50% glycerol and no preservative Human Retinol Binding Recombinant Protein 100μg Each for $797.50
Description & Specifications

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

Retinol Binding Protein (Hu) Recombinant protein RP-75714 detects Retinol Binding Protein (Hu) in Not Applicable samples, and is validated for Control and WB


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 1 mg purified Protein in PBS, with 50% glycerol and no preservative;
Synonyms: RBP

RP-75714 detects Retinol Binding Protein (human) in Not Applicable samples. RP-75714 has been successfully used in Control and Western Blot procedures. RP-75714 The immunogen is N/A.