Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-Resistin (human) Recombinant, Thermo Scientific™

Resistin (Hu) Recombinant protein RP-75711 from Thermo Scientific™ specifically detects Resistin (Hu) in Not Applicable samples, and it is validated for Control and WB

$272.60 - $3,770.00

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human Resistin Recombinant Protein
Recombinant Yes
Source E. coli
View More Specs

 Disclaimers

in vitro research only.

Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

PIRP75709

 
pierce biotechnology inc
RP75709
10μg purified protein in tris HCl with 50% glycerol, 5mM EDTA and no preservative; pH 8 10μg Each for $272.60

PIRP75711

 
pierce biotechnology inc
RP75711
1mg purified protein in PBS with 50% glycerol and no preservative 1mg Each for $3,770.00
Description & Specifications

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human Resistin Recombinant Protein
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

Resistin (Hu) Recombinant protein RP-75711 from Thermo Scientific™ specifically detects Resistin (Hu) in Not Applicable samples, and it is validated for Control and WB


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 1 mg purified Protein in PBS, with 50% glycerol and no preservative;

RP-75711 detects Resistin (human) in Not Applicable samples. RP-75711 has been successfully used in Control and Western Blot procedures. RP-75711 The immunogen is N/A.