Please login to your online account to display your discounted pricing

RecQ DNA Helicase Recombinant Protein, Invitrogen™


RecQ DNA Helicase Recombinant Protein, RP-4903, from Invitrogen™

Manufacturer: pierce biotechnology inc  RP4903

Catalog No. ARC4903

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Control, Western Blot
Formulation 100μg purified protein (0.5mg/mL) in TBS with 0.05M KCl, 50% glycerol, 1mM DTT, 1mM EDTA and no preservative; pH 7.5
Host Species E. Coli
Conjugate Unlabeled
Storage Requirements Store at -80°C, Avoid Freeze/Thaw Cycles
Format Purified
pH Range 7.5
Name RecQ DNA Helicase Recombinant Protein
Purification Method Chromatography
Molecular Weight ∽64
Source E. coli
Concentration 0.5mg/mL
Recombinant Yes
Quantity 100μg

RecQ DNA Helicase Recombinant Protein, RP-4903, from Invitrogen.

E. coli RecQ helicase contributes to the genomic stability as the prototype of RecQ family DNA helicases, mutations of which genes are associated with premature aging and cancer susceptibility, known as Bloom’s and Werner’s syndromes. RecQ helicase functions in both early and late stages of homologous recombination and unwinds DNA with particular structures, such as the fork structure for replication, the Holliday junction which is an intermediate of recombination, telomere, quadruplex G4-DNA, etc. In replication fork, it forms a complex with topoisomerase III and regulates DNA topology.
Host Species: E. Coli
Formulation: 100μg purified protein (0.5mg/mL) in TBS with 0.05M KCl, 50% glycerol, 1mM DTT, 1mM EDTA and no preservative; pH 7.5
RP-4903 can be used as a positive control for Western blots, as well as in studies on mechanisms of DNA recombination, replication and repair. RP-4903 calculated molecular weight is ∽64 kDa.