Please login to your online account to display your discounted pricing

anti-IL-6 (human) Recombinant, Thermo Scientific™


IL-6 (human) Recombinant protein RP-75601 from Thermo Scientific™ specifically detects IL-6 (human) in Not Applicable samples, and it is validated for Control and Western Blot

Manufacturer: pierce biotechnology inc  RP75601

 View more versions of this product

Catalog No. PIRP75601

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Control, Western Blot
Formulation 1mg purified protein in 0.02M tris HCl with 50% glycerol and no preservative; pH 8
Conjugate Unlabeled
Host Species E. Coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Format Purified
Immunogen N/A
Recombinant Yes
Cross Reactivity Human
Name Human IL-6 Recombinant Protein
Quantity 1mg
Source E. coli

IL-6 (human) Recombinant protein RP-75601 from Thermo Scientific™ specifically detects IL-6 (human) in Not Applicable samples, and it is validated for Control and Western Blot


Host Species: E. Coli
Formulation: 1mg purified protein in 0.02M tris HCl with 50% glycerol and no preservative; pH 8
RP-75601 contains purified recombinant human IL-6 protein with an N-terminal hexahistidine tag. RP-75601 may be used as a positive control in many immunological assays.