Please login to your online account to display your discounted pricing

anti-IL-2 (human) Recombinant, Thermo Scientific™


IL-2 (human) Recombinant protein RP-75589 from Thermo Scientific™ specifically detects IL-2 (human) in Not Applicable samples, and it is validated for Control and Western Blot

Manufacturer: pierce biotechnology inc  RP75587

 View more versions of this product

Catalog No. PIRP75587

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
For Use With (Application) Control, Western Blot
Formulation 10μg purified protein (0.44mg/mL) in 20mM tris HCl with 50% glycerol and no preservative; pH 8
Host Species E. Coli
Immunogen N/A
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Format Purified
Name Human IL-2 Recombinant Protein
Quantity 10μg
Recombinant Yes
Cross Reactivity Human
Source E. coli
Concentration 0.05mg/mL

IL-2 (human) Recombinant protein RP-75589 from Thermo Scientific™ specifically detects IL-2 (human) in Not Applicable samples, and it is validated for Control and Western Blot


Host Species: E. Coli
Formulation: 10μg purified protein (0.44mg/mL) in 20mM tris HCl with 50% glycerol and no preservative; pH 8
RP-75587 contains purified recombinant human IL-2 protein with an N-terminal hexahistidine tag. RP-75587 may be used as a positive control in many immunological assays. Purity >90% by SDS-PAGE.