Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-IL-2 (human) Recombinant, Thermo Scientific™

IL-2 (human) Recombinant protein RP-75589 from Thermo Scientific™ specifically detects IL-2 (human) in Not Applicable samples, and it is validated for Control and Western Blot

$326.25 - $4,038.25

Specifications

Cross Reactivity Human
Format Purified
Name Human IL-2 Recombinant Protein
Recombinant Yes
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Source Storage Requirements Price Quantity    

PIRP75589

 
pierce biotechnology inc
RP75589
1mg purified protein in PBS with 50% glycerol and no preservative 1mg E. coli store at -20°C, Avoid Freeze/Thaw Cycles Each for $4,038.25

PIRP75587

 
pierce biotechnology inc
RP75587
10μg purified protein (0.44mg/mL) in 20mM tris HCl with 50% glycerol and no preservative; pH 8 10μg E. coli store at -20°C, Avoid Freeze/Thaw Cycles Each for $326.25

PIRP75588

 
pierce biotechnology inc
RP75588
100μg purified protein in tris HCl with 50% glycerol and no preservative; pH 8 100μg E. coli store at -20°C, Avoid Freeze/Thaw Cycles Each for $785.90
Description & Specifications

Specifications

Cross Reactivity Human
Format Purified
Name Human IL-2 Recombinant Protein
Recombinant Yes

IL-2 (human) Recombinant protein RP-75589 from Thermo Scientific™ specifically detects IL-2 (human) in Not Applicable samples, and it is validated for Control and Western Blot


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 1 mg purified Protein in PBS, with 50% glycerol and no preservative;

RP-75589 detects IL-2 (human) in Not Applicable samples. RP-75589 has been successfully used in Control and Western Blot procedures. RP-75589 The immunogen is N/A.