Please login to your online account to display your discounted pricing

IL-2 (human) Recombinant Protein, Invitrogen™


IL-2 (human) Recombinant Protein, RP-75588, from Invitrogen™

Manufacturer: pierce biotechnology inc  RP75588

Catalog No. PIRP75588

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Control, Western Blot
Format Purified
Formulation 100μg purified protein in tris HCl with 50% glycerol and no preservative; pH 8
Name Human IL-2 Recombinant Protein
pH Range 8
Quantity 100μg
Source E. coli
Immunogen N/A
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Conjugate Unlabeled
Cross Reactivity Human
Host Species E. coli
Recombinant Yes

IL-2 (human) Recombinant Protein, RP-75588, from Invitrogen.


Host Species: E. Coli
Formulation: 100μg purified protein in tris HCl with 50% glycerol and no preservative; pH 8
RP-75588 contains purified recombinant human IL-2 protein with an N-terminal hexahistidine tag. RP-75588 may be used as a positive control in many immunological assays.