Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-IL-10 (human) Recombinant, Thermo Scientific™

IL-10 (human) Recombinant protein RP-75613 from Thermo Scientific™ specifically detects IL-10 (human) in Not Applicable samples, and it is validated for Control and Western Blot

$268.25 - $3,770.00

Specifications

Cross Reactivity Human
Name Human IL-10 Recombinant Protein
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
View More Specs
Products
Catalog Number Mfr. No. For Use With (Application) Formulation Quantity pH Range Price Quantity    

PIRP75611

 
pierce biotechnology inc
RP75611
Control, Western Blot 10μg purified protein (0.10mg/mL) in tris with 50% glycerol and no preservative; pH 8 10μg 8 Each for $268.25

PIRP75612

 
pierce biotechnology inc
RP75612
Control, Western Blot 100μg purified protein (0.25mg/mL) in 0.01M tris HCl with 50% glycerol, 0.25M NaCl and no preservative; pH 8 100μg 8 Each for $759.80

PIRP75613

 
pierce biotechnology inc
RP75613
Control, Western Blot 1mg purified protein in 0.02M tris with 50% glycerol and no preservative; pH 8 1mg 8 Each for $3,770.00
Description & Specifications

Specifications

Cross Reactivity Human
Name Human IL-10 Recombinant Protein
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

IL-10 (human) Recombinant protein RP-75613 from Thermo Scientific™ specifically detects IL-10 (human) in Not Applicable samples, and it is validated for Control and Western Blot


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 1 mg purified Protein in 0.02M tris, with 50% glycerol and no preservative; 8

RP-75613 detects IL-10 (human) in Not Applicable samples. RP-75613 has been successfully used in Control and Western Blot procedures. RP-75613 The immunogen is N/A.