Please login to your online account to display your discounted pricing

anti-RAD52 (human) Recombinant, Thermo Scientific™


RAD52 (human) Recombinant protein RP-4973 from Thermo Scientific™ specifically detects RAD52 (human) in Not Applicable samples, and it is validated for Control and Western Blot

Manufacturer: pierce biotechnology inc  RP4972

 View more versions of this product

Catalog No. PIRP4972

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
For Use With (Application) Control, Western Blot
Formulation 20μg purified protein (1mg/mL) in 0.02M potassium phosphate with 0.2M KCl, 20% glycerol, 2mM beta mercaptoethanol, 0.5mM EDTA and no preservative; pH 7.5
Host Species E. coli
Storage Requirements Store at -80°C, Avoid Freeze/Thaw Cycles
Format Purified
Name Human RAD52 Recombinant Protein
pH Range 7.5
Quantity 20μg
Cross Reactivity Human
Concentration 1mg/mL
Recombinant Yes
Source E. coli

RAD52 (human) Recombinant protein RP-4973 from Thermo Scientific™ specifically detects RAD52 (human) in Not Applicable samples, and it is validated for Control and Western Blot

Human Rad52 protein plays a major role in genetic recombination and recombination repair by mediating strand-annealing reaction between complementary DNA strands. Rad52 functionally and physically interacts with Rad51, Dmc1, and RPA in recombination processes. In radiation damaged cell, Rad52 is phosphorylated at Tyr104 by c-Abl tyrosine kinase and forms nuclear foci.
Host Species: E. coli
Formulation: 20μg purified protein (1mg/mL) in 0.02M potassium phosphate with 0.2M KCl, 20% glycerol, 2mM beta mercaptoethanol, 0.5mM EDTA and no preservative; pH 7.5
RP-4972 contains highly purified recombinant Rad52 protein expressed in E. coli. RP-4972 may be used as a control in Western blotting applications.