Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-RAD52 (human) Recombinant, Thermo Scientific™

RAD52 (human) Recombinant protein RP-4973 from Thermo Scientific™ specifically detects RAD52 (human) in Not Applicable samples, and it is validated for Control and Western Blot

$1,142.60 - $2,755.00

Specifications

Concentration 1mg/mL
Cross Reactivity Human
Formulation 20μg purified protein (1mg/mL) in 0.02M potassium phosphate with 0.2M KCl, 20% glycerol, 2mM beta mercaptoethanol, 0.5mM EDTA and no preservative; pH 7.5
For Use With (Application) Control, Western Blot
Name Human RAD52 Recombinant Protein
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

PIRP4973

 
pierce biotechnology inc
RP4973
100μg Each for $2,755.00

PIRP4972

 
pierce biotechnology inc
RP4972
20μg Each for $1,142.60
Description & Specifications

Specifications

Concentration 1mg/mL
Cross Reactivity Human
Formulation 20μg purified protein (1mg/mL) in 0.02M potassium phosphate with 0.2M KCl, 20% glycerol, 2mM beta mercaptoethanol, 0.5mM EDTA and no preservative; pH 7.5
For Use With (Application) Control, Western Blot
Name Human RAD52 Recombinant Protein
pH Range 7.5
Recombinant Yes
Source E. coli
Storage Requirements Store at -80°C, Avoid Freeze/Thaw Cycles

RAD52 (human) Recombinant protein RP-4973 from Thermo Scientific™ specifically detects RAD52 (human) in Not Applicable samples, and it is validated for Control and Western Blot

Human Rad52 protein plays a major role in genetic recombination and recombination repair by mediating strand-annealing reaction between complementary DNA strands. Rad52 functionally and physically interacts with Rad51, Dmc1, and RPA in recombination processes. In radiation damaged cell, Rad52 is phosphorylated at Tyr104 by c-Abl tyrosine kinase and forms nuclear foci.
Host Species: Human
Cross Reactivity: Not Applicable
Formulation: 100μg purified Protein (1mg/mL) in 0.02M potassium phosphate, with 0.2M KCl, 20% glycerol, 2mM beta mercaptoethanol, 0.5mM EDTA and no preservative; 7.5

RP-4973 detects RAD52 (human) in Not Applicable samples. RP-4973 has been successfully used in Control and Western Blot procedures. RP-4973