Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-RAD51 Recombinant, Thermo Scientific™

RAD51 Recombinant protein RP-4900 from Thermo Scientific™ specifically detects RAD51 in Not Applicable samples, and it is validated for Control and Western Blot

$1,142.60 - $1,142.60

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human Rad51 Recombinant Protein
pH Range 8
Recombinant Yes
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    

ARC4964

 
pierce biotechnology inc
RP4964
Each for $1,142.60
Description & Specifications

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human Rad51 Recombinant Protein
pH Range 8
Recombinant Yes
Source E. coli

RAD51 Recombinant protein RP-4900 from Thermo Scientific™ specifically detects RAD51 in Not Applicable samples, and it is validated for Control and Western Blot

E. coli RecA protein is a very important enzyme for homologous recombination and recombinational repair. Expression is induced by SOS response caused by DNA damage. The RecA protein has multiple functions, such as single stranded DNA dependent ATPase activity, DNA annealing activity, formation of D-loops and Holliday structures during homologous recombination, and co-protease activities that promote self-cleavage of the LexA repressor, lambda phage repressor and UmuD protein. RecA protein binds to single and double stranded DNA for nucleofilament formation. It carries out a central role in homologous recombination. Its is homologous to the eukaryotic Rad51 protein and Dmcl protein.
Host Species: E. Coli
Cross Reactivity: Not Applicable
Formulation: 100μg purified Protein (0.9mg/mL) in TBS, with 0.15M KCl, 50% glycerol, 1mM DTT, 1mM EDTA and no preservative; 8.0
Synonyms: RAD51A

RP-4900 detects RAD51 in Not Applicable samples. RP-4900 has been successfully used in Control and Western Blot procedures. RP-4900