Please login to your online account to display your discounted pricing

anti-RAD51 Recombinant, Thermo Scientific™


RAD51 Recombinant protein RP-4900 from Thermo Scientific™ specifically detects RAD51 in Not Applicable samples, and it is validated for Control and Western Blot

Manufacturer: pierce biotechnology inc  RP4964

 View more versions of this product

Catalog No. ARC4964

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
For Use With (Application) Control, Western Blot
Formulation 20μg purified protein (1mg/mL) in tris HCl with 0.01M KCl, 10% glycerol, 1mM DTT, 0.5mM EDTA and no preservative; pH 8
Host Species E. Coli
Storage Requirements Store at -80°C, Avoid Freeze/Thaw Cycles
Gene Alias RAD51 homolog
Format Purified
Molecular Weight ∽35
Name Human Rad51 Recombinant Protein
pH Range 8
Quantity 20μg
Recombinant Yes
Source E. coli
Concentration 1mg/mL
Cross Reactivity Human

RAD51 Recombinant protein RP-4900 from Thermo Scientific™ specifically detects RAD51 in Not Applicable samples, and it is validated for Control and Western Blot

Human Rad51 protein is a functional and structural homolog of E. coli RecA protein, which plays a major role in genetic recombination and recombination repair by mediating strand exchange reaction between homologous DNA strands. Rad51 functionally and physically interacts with its paralogs Dmc1, Rad51B, Rad51C, Rad51D, Xrcc2 and Xrcc3, and also with Rad52 in recombination processes. It also interacts with oncogene proteins and tumor suppressors such as BRCA1, BRCA2, and P53 for the maintenance of genome stability.
Host Species: E. Coli
Formulation: 20μg purified protein (1mg/mL) in tris HCl with 0.01M KCl, 10% glycerol, 1mM DTT, 0.5mM EDTA and no preservative; pH 8
RP-4964 has been successfully used in Western blot proceedures. It is specific for the ∽35 kDa Rad51 protein. The Human gene Rad51 is homologous to the highly conserved bacterial gene RecA. RP-4964 is highly purified from E. coli over-expressing human Rad51 protein as a recombinant protein. The tag was removed from the recombinant protein (though it still contains Gly-Ser-His at the N-terminal). The protein has been shown to retain nuclear filament forming and strand-exchange activity as well as interaction with Rad52. RP-4964 was confirmed to possess single strand DNA stimulated ATPase activity.