Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-Procalcitonin (human) Recombinant, Thermo Scientific™

Procalcitonin (Hu) Recombinant protein RP-75699 from Thermo Scientific™ specifically detects Procalcitonin (Hu) in Not Applicable samples, and it is validated for Control and WB

$723.55 - $4,371.75

Specifications

Concentration 0.89mg/mL
Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human Procalcitonin Recombinant Protein
pH Range 8
View More Specs

 Disclaimers

in vitro research only.

Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

PIRP75698

 
pierce biotechnology inc
RP75698
100μg purified protein (0.72mg/mL) in tris HCl with 50% glycerol and no preservative; pH 8 100μg Each for $723.55

PIRP75699

 
pierce biotechnology inc
RP75699
1mg purified protein (1mg/mL) in tris HCl with 50% glycerol and no preservative; pH 8.0 1mg Each for $4,371.75
Description & Specifications

Specifications

Concentration 0.89mg/mL
Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human Procalcitonin Recombinant Protein
pH Range 8
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

Procalcitonin (Hu) Recombinant protein RP-75699 from Thermo Scientific™ specifically detects Procalcitonin (Hu) in Not Applicable samples, and it is validated for Control and WB


Host Species: E. Coli
Cross Reactivity: Not Applicable
Formulation: 1 mg purified Protein (1mg/mL) in tris HCl, with 50% glycerol and no preservative; 8.0
Synonyms: Calcitonin

RP-75699 detects Procalcitonin (human) in Not Applicable samples. RP-75699 has been successfully used in Control and Western Blot procedures. RP-75699