Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-Pregnancy Assoc. Plasma Protein-1 (human) Recombinant, Thermo Scientific™

Pregnancy Assoc. Plasma Protein-1 (Hu) Recombinant protein RP-75696 detects Pregnancy Assoc. Plasma Protein-1 (Hu) in Not Applicable samples, and is validated for Control and WB

$288.55 - $3,770.00

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human Pregnancy Associated Plasma Protein-1 Recombinant Protein
Recombinant Yes
Source E. coli
View More Specs

 Disclaimers

in vitro research only.

Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

PIRP75694

 
pierce biotechnology inc
RP75694
10μg purified protein in PBS with 50% glycerol and no preservative 10μg Each for $288.55

PIRP75695

 
pierce biotechnology inc
RP75695
100μg purified protein in PBS with 50% glycerol and no preservative 100μg Each for $797.50

PIRP75696

 
pierce biotechnology inc
RP75696
1mg purified protein in PBS with 50% glycerol and no preservative 1mg Each for $3,770.00
Description & Specifications

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human Pregnancy Associated Plasma Protein-1 Recombinant Protein
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

Pregnancy Assoc. Plasma Protein-1 (Hu) Recombinant protein RP-75696 detects Pregnancy Assoc. Plasma Protein-1 (Hu) in Not Applicable samples, and is validated for Control and WB


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 1 mg purified Protein in PBS, with 50% glycerol and no preservative;
Synonyms: PAPPA1

RP-75696 detects Pregnancy Assoc. Plasma Protein-1 (human) in Not Applicable samples. RP-75696 has been successfully used in Control and Western Blot procedures. RP-75696 The immunogen is N/A.