Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-Plasminogen Activator Inhibitor 1 (human) Recombinant, Thermo Scientific™

Plasminogen Activator Inhibitor 1 (Hu) Recombinant protein RP-75687 detects Plasminogen Activator Inhibitor 1 (Hu) in Not Applicable samples, and is validated for Control and WB

$288.55 - $3,770.00

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human Plasminogen Activator Inhibitor 1 Recombinant Protein
Recombinant Yes
Source E. coli
View More Specs

 Disclaimers

in vitro research only.

Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

PIRP75687

 
pierce biotechnology inc
RP75687
1mg purified protein in PBS with 50% glycerol and no preservative 1mg Each for $3,770.00

PIRP75685

 
pierce biotechnology inc
RP75685
10μg purified protein in PBS with 50% glycerol and no preservative 10μg Each for $288.55

PIRP75686

 
pierce biotechnology inc
RP75686
100μg purified protein (0.12mg/mL) in 0.01M Bis-tris propane with 0.01M NaCl, 50% glycerol and no preservative; pH 9 100μg Each for $797.50
Description & Specifications

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human Plasminogen Activator Inhibitor 1 Recombinant Protein
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

Plasminogen Activator Inhibitor 1 (Hu) Recombinant protein RP-75687 detects Plasminogen Activator Inhibitor 1 (Hu) in Not Applicable samples, and is validated for Control and WB


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 1 mg purified Protein in PBS, with 50% glycerol and no preservative;
Synonyms: PAI1

RP-75687 detects Plasminogen Activator Inhibitor 1 (human) in Not Applicable samples. RP-75687 has been successfully used in Control and Western Blot procedures. RP-75687 The immunogen is N/A.