Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-PKM2 Pyruvate Kinase (human) Recombinant, Thermo Scientific™

PKM2 Pyruvate Kinase (Hu) Recombinant protein RP-75744 from Thermo Scientific™ detects PKM2 Pyruvate Kinase (Hu) in Not Applicable samples, and is validated for Control and WB

$326.25 - $3,770.00

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human PKM2 Pyruvate Kinase Recombinant Protein
Recombinant Yes
Source E. coli
View More Specs

 Disclaimers

in vitro research only.

Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

PIRP75744

 
pierce biotechnology inc
RP75744
1mg purified protein in PBS with 50% glycerol and no preservative 1mg Each for $3,770.00

PIRP75743

 
pierce biotechnology inc
RP75743
100μg purified protein in tris HCl with 50% glycerol and no preservative; pH 8 100μg Each for $759.80

PIRP75742

 
pierce biotechnology inc
RP75742
10μg purified protein in 20mM tris HCl with 50% glycerol and no preservative; pH 8.0 10μg Each for $326.25
Description & Specifications

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human PKM2 Pyruvate Kinase Recombinant Protein
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

PKM2 Pyruvate Kinase (Hu) Recombinant protein RP-75744 from Thermo Scientific™ detects PKM2 Pyruvate Kinase (Hu) in Not Applicable samples, and is validated for Control and WB


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 1 mg purified Protein in PBS, with 50% glycerol and no preservative;
Synonyms: Tumor Type M2 Pyruvate Kinase

RP-75744 detects PKM2 Pyruvate Kinase (human) in Not Applicable samples. RP-75744 has been successfully used in Control and Western Blot procedures. RP-75744 The immunogen is N/A.