Please login to your online account to display your discounted pricing

anti-Phalloidin , Thermo Scientific™

Phalloidin control 21833 from Thermo Scientific™ specifically detects Phalloidin in Many samples, and it is validated for ICC, IF and IHC

$527.10 - $527.10

Specifications

Cross Reactivity Many Species
Format Purified
Formulation 300 units purified control antibody (300 units/ml) in methanol and no preservative
Storage Requirements Store at -20°C
View More Specs
Products
Catalog Number Mfr. No. Conjugate Price Quantity    

PI21836

 
pierce biotechnology inc
21836
DyLight 594 Each for $527.10
Description & Specifications

Specifications

Cross Reactivity Many Species
Format Purified
Formulation 300 units purified control antibody (300 units/ml) in methanol and no preservative
Storage Requirements Store at -20°C

Phalloidin control 21833 from Thermo Scientific™ specifically detects Phalloidin in Many samples, and it is validated for ICC, IF and IHC

CCR4 is a chemokine receptor and is preferentially expressed on type 2 helper T (Th2-type) cells. In contrast to other chemokine receptors, the expression of CCR4 and CCR8 on Th2 cells is transiently increased following TCR and CD28 engagement. Moreover, activated Th1 cells up-regulate CCR4 expression and functional responsiveness to thymus- and activation-regulated chemokine.
Cross Reactivity: Many
Formulation: 300 units purified Control (300 units/ml) in methanol, with no preservative;
Synonyms: DY488-Phalloidin, F-actin toxin, Amanita phalloides

PA5-32698 detects CCR4 in Human samples. PA5-32698 has been successfully used in ELISA and Immunohistochemistry (Paraffin) procedures. PA5-32698 The immunogen is Synthetic 18 amino acid peptide from 2nd extracellular domain of human CCR4..