missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phalloidin DyLight 550, Invitrogen™

Phalloidin Control, DyLight 550 conjugate, 21835, from Invitrogen™., 21835, from Invitrogen™. Species Reactivity: Many; Applications: ICC, IF, IHC.

Manufacturer:  Pierce Biotechnology Inc 21835*

Catalog No. PI21835


 

Specifications

Description

Phalloidin Control, DyLight 550 conjugate, 21835, from Invitrogen., 21835, from Invitrogen. Species Reactivity: Many; Applications: Immunocytochemistry, Immunofluorescence, Immunohistochemistry.

Phalloidin is a bicyclic peptide that belongs to a family of toxins isolated from the deadly Amanita phalloides “death cap” mushroom and is commonly used as a counterstain (similar to DAPI or Hoechst) in cell biology and histology imaging applications to selectively label F-actin in fixed cells, permeabilized cells, and cell-free experiments. Labeled phalloidin conjugates have similar affinity for both large and small filaments and bind in a stoichiometric ratio of about one phallotoxin per actin subunit in both muscle and non-muscle cells. Phalloidins reportedly do not bind to monomeric G-actin, unlike some antibodies against actin. The dynamics of the actin polymerization in cells are important for a variety of cellular processes from cell motility to cell shape, from muscular contraction to cytokinesis, and more.
Cross Reactivity: Many Species
Formulation: 300 units purified control antibody (300 units/ml) in methanol and no preservative
Synonyms: DY550-Phalloidin, F-actin toxin, Amanita phalloides
Format: 300 units of lyophilized DyLight 550-Phalloidin (300 units/ml) in methanol. Prepare stock solution by adding 1000µl of pure methanol to the vial and gently mix. 21835 has been successfully used in immunofluorescence and immunohistochemistry. DyLight 550-Phalloidin has an excitation/emission of 557/573nm and molecular weight of 1452.68 g/mole. DyLight 550-Phalloidin Stock Solution is prepared in methanol, and is also soluble in DMF, or DMSO. The unused Stock Solution should be promptly stored at -20°C in a foil pouch with desiccant, and protected from light. The stock solutions are stable for at least one year. Staining cells with some of the DyLight Phalloidin conjugates may require the use of a higher concentration of the phalloidin conjugate; therefore to minimize the amount of methanol added to the cells, the vial contents could be dissolved in 0.5mL methanol to yield a final concentration of 600units/mL. Working solution of 1unit/mL (per 96-well plate) can be prepared by diluting 20µl of the Stock Solution in 5.98mL of PBS and mixing well. Typical staining procedure adds 50µl of Working Solution (i.e. 1 to 5 unit/ml) to a each well. Incubate cells in the dark for 30 minutes at room temperature (optimal staining times varies from 10 minutes to 3 hours depending on cell type). Aspirate and wash cells three times in PBS after incubation. If desired, probe with specific primary antibodies followed by secondary antibodies conjugated to any compatible fluorophores before using DyLight 550-Phalloidin conjugate.
Description & Specifications

Specifications

Phalloidin
DyLight 550
Purified
300U
Many
Immunocytochemistry,Immunofluorescence,Immunohistochemistry
300 units purified control antibody (300 units/ml) in methanol and no preservative
DY550-Phalloidin, F-actin toxin, Amanita phalloides
Store at -20°C