Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-PPAR gamma (human) Recombinant, Thermo Scientific™

PPAR gamma (Hu) Recombinant protein RP-75684 from Thermo Scientific™ specifically detects PPAR gamma (Hu) in Not Applicable samples, and it is validated for Control and WB

$319.00 - $4,038.25

Specifications

Concentration 0.27mg/mL
Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human PPAR gamma Recombinant Protein
pH Range 8
View More Specs

 Disclaimers

in vitro research only.

Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

PIRP75682

 
pierce biotechnology inc
RP75682
10μg purified protein (0.32mg/mL) in 0.025M tris HCl with 50% glycerol, 0.01mM EDTA, 75mM NaCl and no preservative; pH 8.0 10μg Each for $319.00

PIRP75684

 
pierce biotechnology inc
RP75684
1mg purified protein in 20mM tris HCl with 50% glycerol and no preservative; pH 8 1mg Each for $4,038.25
Description & Specifications

Specifications

Concentration 0.27mg/mL
Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human PPAR gamma Recombinant Protein
pH Range 8
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

PPAR gamma (Hu) Recombinant protein RP-75684 from Thermo Scientific™ specifically detects PPAR gamma (Hu) in Not Applicable samples, and it is validated for Control and WB


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 1 mg purified Protein in 20mM tris HCl, with 50% glycerol and no preservative; 8
Synonyms: Peroxisome Proliferator-Acivated Receptor Gamma

RP-75684 detects PPAR gamma (human) in Not Applicable samples. RP-75684 has been successfully used in Control and Western Blot procedures. RP-75684 The immunogen is N/A.