Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-PDGF B (human) Recombinant, Thermo Scientific™

PDGF B (Hu) Recombinant protein RP-75693 from Thermo Scientific™ specifically detects PDGF B (Hu) in Not Applicable samples, and it is validated for Control and WB

$319.00 - $3,770.00

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human PDGF B Recombinant Protein
pH Range 4.8
Recombinant Yes
View More Specs

 Disclaimers

in vitro research only.

Products
Catalog Number Mfr. No. Concentration Formulation Quantity Price Quantity    

PIRP75691

 
pierce biotechnology inc
RP75691
0.16mg/mL 10μg purified protein (0.1mg/mL) in 0.025M acetic acid with 1mM EDTA, 50% glycerol and no preservative; pH 4.8 10μg Each for $319.00

PIRP75692

 
pierce biotechnology inc
RP75692
0.1mg/mL 100μg purified protein in 0.025M acetic acid with 50% glycerol, 1mM EDTA and no preservative; pH 4.8 100μg Each for $785.90

PIRP75693

 
pierce biotechnology inc
RP75693
0.16mg/mL 1mg purified protein in PBS with 50% glycerol and no preservative 1mg Each for $3,770.00
Description & Specifications

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human PDGF B Recombinant Protein
pH Range 4.8
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

PDGF B (Hu) Recombinant protein RP-75693 from Thermo Scientific™ specifically detects PDGF B (Hu) in Not Applicable samples, and it is validated for Control and WB


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 1 mg purified Protein in PBS, with 50% glycerol and no preservative;

RP-75693 detects PDGF B (human) in Not Applicable samples. RP-75693 has been successfully used in Control and Western Blot procedures. RP-75693 The immunogen is N/A.