Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-Parathyroid Hormone Recombinant, Thermo Scientific™

Parathyroid Hormone Recombinant protein RP-10926 from Thermo Scientific™ specifically detects Parathyroid Hormone in Not Applicable samples, and it is validated for Control

$509.25 - $955.50

Specifications

Format Purified
For Use With (Application) Control
Name Parathyroid Hormone Recombinant Protein
Recombinant Yes
Source E. coli
View More Specs

 Disclaimers

in vitro research only.

Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

PIRP10926

 
pierce biotechnology inc
RP10926
100μg purified protein in citric acid with 0.1117mg EDTA, 50mg mannitol, 0.2mg Tween 80 and no preservative 100μg Each for $509.25

PIRP10937

 
pierce biotechnology inc
RP10937
500μg purified protein in citric acid with 0.1117mg EDTA, 50mg mannitol, 0.2mg Tween 80 and no preservative 500μg Each for $955.50
Description & Specifications

Specifications

Format Purified
For Use With (Application) Control
Name Parathyroid Hormone Recombinant Protein
Recombinant Yes
Source E. coli
Storage Requirements Store at -80°C, Avoid Freeze/Thaw Cycles

Parathyroid Hormone Recombinant protein RP-10926 from Thermo Scientific™ specifically detects Parathyroid Hormone in Not Applicable samples, and it is validated for Control


Host Species: E. coli
Cross Reactivity: Not Applicable
Clone: N/A
Immunogen: N/A
Formulation: 100μg purified Protein in citric acid, with 0.1117mg EDTA, 50mg mannitol, 0.2mg Tween 80 and no preservative;
Synonyms: PTH

RP-10926 detects Parathyroid Hormone in Not Applicable samples. RP-10926 has been successfully used in Control procedures. RP-10926 The immunogen is N/A.