Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-Paraoxonase 2 (human) Recombinant, Thermo Scientific™

Paraoxonase 2 (Hu) Recombinant protein RP-75681 from Thermo Scientific™ specifically detects Paraoxonase 2 (Hu) in Not Applicable samples, and it is validated for Control and WB

$288.55 - $3,770.00

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human Paraoxonase 2 Recombinant Protein
Recombinant Yes
Source E. coli
View More Specs

 Disclaimers

in vitro research only.

Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

PIRP75680

 
pierce biotechnology inc
RP75680
100μg purified protein in 20mM tris HCl with 50% glycerol and no preservative 100μg Each for $797.50

PIRP75679

 
pierce biotechnology inc
RP75679
10μg purified protein in PBS with 50% glycerol and no preservative 10μg Each for $288.55

PIRP75681

 
pierce biotechnology inc
RP75681
1mg purified protein in PBS with 50% glycerol and no preservative 1mg Each for $3,770.00
Description & Specifications

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human Paraoxonase 2 Recombinant Protein
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

Paraoxonase 2 (Hu) Recombinant protein RP-75681 from Thermo Scientific™ specifically detects Paraoxonase 2 (Hu) in Not Applicable samples, and it is validated for Control and WB


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 1 mg purified Protein in PBS, with 50% glycerol and no preservative;

RP-75681 detects Paraoxonase 2 (human) in Not Applicable samples. RP-75681 has been successfully used in Control and Western Blot procedures. RP-75681 The immunogen is N/A.