Please login to your online account to display your discounted pricing

Mutant p53 (human) Recombinant, Invitrogen™


mutant p53 (Hu) Recombinant protein RP-820 detects mutant p53 (Hu) in Not Applicable samples, and is validated for Control, ELISA, Inhibition Assays and WB

Manufacturer: pierce biotechnology inc  RP820

Catalog No. ARC820

  • / Each
 

Description & Specifications

Specifications

Storage Requirements store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Formulation 5μg purified Protein in sodium phosphate, 0.01M tris, with 8M urea and no preservative;
Format Purified
Source E. coli
For Use With (Application) Control (Assay Dependent), ELISA, Inhibition Assay, Western Blotting
Purification Method Chromatography
Molecular Weight ∽53
Name Human Mutant p53 Recombinant Protein
Recombinant Yes
Quantity 5μg
Cross Reactivity Human

mutant p53 (Hu) Recombinant protein RP-820 detects mutant p53 (Hu) in Not Applicable samples, and is validated for Control, ELISA, Inhibition Assays and WB

p53 is a tumor-suppressor gene expressed in a wide variety of tissue types and is involved in regulating cell growth, replication, and apoptosis. The p53 protein (393 aa, phosphoprotein) is a transcription factor, whose activity is regulated by phos-phorylation. The normal function of p53 is to effect cell cycle arrest at the G1- and G2-checkpoints in response to DNA damage. The current model postulates that p53 senses DNA damage and arrests the cell cycle to allow DNA repair to take place. If repair is not successful, p53 initiates programmed cell death. In normal cells the p53 level is low. In response to DNA damage or other cellular stress signals the checkpoint function is executed by accumulation of p53. The control function of p53 includes the transcriptional regulation of several genes including the cell cycle inhibitor p21 WAF1/CIP1/SDI1 , DNA repair gene GADD45 and the apoptotic inducer Bax. p53 has also been shown to induce apoptosis by means of a direct signaling pathway. p53 directly binds to and acts on several cellular proteins involved in various pathways, including c-Abl or basal transcription factors. p53 can be functionally inactivated by mutation, most of them being amino acid substitutions. Inactivation can also be the result of binding to DNA tumor virus encoded proteins such as SV40 large T antigen, adenovirus E1B- and papilloma Virus E6-proteins or as a consequence of its interaction with the oncogene-encoded protein MDM2. The p53 tumor suppressor gene has been found to contain mutation in over 50% of human cancers. Many mutations of the p53 gene have been associated with malignant transformation in a wide variety of human tumors, including breast, ovarian, bladder, colon, lung, and melanoma.
Cross Reactivity: Not Applicable
Formulation: 5μg purified Protein in sodium phosphate, 0.01M tris, with 8M urea and no preservative;

RP-820 detects mutant p53 (human) in Not Applicable samples. RP-820 has been successfully used in Control, ELISA, Inhibition Assays and Western Blot procedures. RP-820