Please login to your online account to display your discounted pricing

P-Selectin (human) Recombinant, Invitrogen™


P-Selectin (Hu) Recombinant protein RP-75705 from Thermo Scientific™ specifically detects P-Selectin (Hu) in Not Applicable samples, and it is validated for Control and WB

Manufacturer: pierce biotechnology inc  RP75705

Catalog No. PIRP75705

  • / Each
 

Description & Specifications

Specifications

Immunogen N/A
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Format Purified
Formulation 1 mg purified Protein in 25mM acetic acid, with 5mM EDTA, 50% glycerol and no preservative; 4.8
pH Range 4.8
Molecular Weight 65.3
Name Human P-Selectin Recombinant Protein
Quantity 1mg
Recombinant Yes
Source E. coli
Cross Reactivity Human
For Use With (Application) Control (Assay Dependent), Western Blotting

P-Selectin (Hu) Recombinant protein RP-75705 from Thermo Scientific™ specifically detects P-Selectin (Hu) in Not Applicable samples, and it is validated for Control and WB


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 1 mg purified Protein in 25mM acetic acid, with 5mM EDTA, 50% glycerol and no preservative; 4.8

RP-75705 detects P-Selectin (human) in Not Applicable samples. RP-75705 has been successfully used in Control and Western Blot procedures. RP-75705 The immunogen is N/A.