Please login to your online account to display your discounted pricing

anti-Rat AA-NAT (residues 198-208) Synthetic, Thermo Scientific™


Rt AA-NAT (residues 198-208) Synthetic peptide PEP-065 from Thermo Scientific™ detects Rt AA-NAT (residues 198-208) in Not Applicable samples, and is validated for Control

Manufacturer: pierce biotechnology inc  PEP065

 View more versions of this product

Catalog No. PEP065

  • / Each
 

Description & Specifications

Specifications

Format Purified
Formulation 50μg purified peptide (0.5mg/mL) and no preservative
For Use With (Application) Control
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Product Type Rat AA-NAT (Residues 198-208) Synthetic Peptide
Lyophilized Yes
Purity >95%
Sequence C198-H-T-F-L-R-R-N-S-G-C208
Concentration 0.5mg/mL
Quantity 50μg

Rt AA-NAT (residues 198-208) Synthetic peptide PEP-065 from Thermo Scientific™ detects Rt AA-NAT (residues 198-208) in Not Applicable samples, and is validated for Control


Formulation: 50μg purified peptide (0.5mg/mL) and no preservative
PEP-065 is a 11 amino acid synthetic peptide whose sequence corresponds to residues 198-208 of rat AA-NAT. The sequence of this peptide is (amino to carboxy terminus): C198-H-T-F-L-R-R-N-S-G-C208 This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and AA-NAT, catalog # PA1-762. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∽70-fold) for competitive inhibition of antibody-protein binding reactions. Reconstitute with 0.1 ml of distilled water.