Please login to your online account to display your discounted pricing

anti-Rat AA-NAT (residues 198-208) Synthetic, Thermo Scientific™


Rt AA-NAT (residues 198-208) Synthetic peptide PEP-065 from Thermo Scientific™ detects Rt AA-NAT (residues 198-208) in Not Applicable samples, and is validated for Control

Manufacturer: pierce biotechnology inc  PEP063

 View more versions of this product

Catalog No. PEP063

  • / Each
 

Description & Specifications

Specifications

Format Purified
For Use With (Application) Control
Formulation 50μg purified peptide (0.5mg/mL) and no preservative
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Product Type Rat AA-Nat (Residues 24-33) Synthetic Peptide
Lyophilized Yes
Purity >95%
Quantity 50μg
Concentration 0.5mg/mL
Sequence C24-Q-R-R-H-T-L-P-A-S33

Rt AA-NAT (residues 198-208) Synthetic peptide PEP-065 from Thermo Scientific™ detects Rt AA-NAT (residues 198-208) in Not Applicable samples, and is validated for Control


Formulation: 50μg purified peptide (0.5mg/mL) and no preservative
PEP-063 is a 10 amino acid synthetic peptide whose sequence corresponds to residues 24-33 of rat AA-NAT. The sequence of this peptide is (amino to carboxy terminus): C24-Q-R-R-H-T-L-P-A-S33 This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and AA-NAT, catalog # PA1-760. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∽70-fold) for competitive inhibition of antibody-protein binding reactions. Reconstitute with 0.1 ml of distilled water.